Learn More
Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector
Made from Kynar. Ark-Plas Products™ Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector is ideal for connecting tubes of various diameter. The connector offers easy fitting into tubing or hoses without the need for tools.
Brand:  Ark-Plas Products™ MAD60-DF0
 View more versions of this product
Product Code. 12323850
  • 183.00 NOK / Pack of 10
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
Description
  • Made of Kynar
  • Economical straight barbed
  • Easily connect fluid paths without the use of tools
Specifications
Specifications
Straight reducing connector
Fluororesin
3/16 x 1/16 in.
Natural