missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU2AF1 Mouse anti-Human, Clone: 3C5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66659-1-IG-20UL
This item is not returnable.
View return policy
Description
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.Specifications
POU2AF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16633 | |
POU2AF1 | |
POU2AF1 Fusion Protein Ag23613 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
3C5A7 | |
Unconjugated | |
POU2AF1 | |
BOB 1, BOB1, OBF 1, OBF1, OCA B, OCAB, OCT binding factor 1, POU2AF1 | |
Mouse | |
Protein G | |
RUO | |
5450 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction