missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU2AF1 Mouse anti-Human, Clone: 3C5A7, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | POU2AF1 |
---|---|
Clone | 3C5A7 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16848745
|
Proteintech
66659-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16838745
|
Proteintech
66659-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.Specifications
POU2AF1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BOB 1, BOB1, OBF 1, OBF1, OCA B, OCAB, OCT binding factor 1, POU2AF1 | |
POU2AF1 | |
IgG1 | |
Protein G | |
Antibody |
3C5A7 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q16633 | |
5450 | |
POU2AF1 Fusion Protein Ag23613 | |
Primary | |
-20°C | |
POU2AF1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title