missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Potassium-Sodium Diet, Low, Pellet

Diet low in sodium (Na) and potassium (K).

7995.58 NOK - 30979.56 NOK

Specifications

Product Type Animal Diet
Autoclavable Autoclavable
Disposable Single-use
Format Pellet
Sterility Non-sterile
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443031
View Documents
MP Biomedicals
0296023010
10 kg
7995.58 NOK
10kg
Please to purchase this item. Need a web account? Register with us today!
11457091
View Documents
MP Biomedicals
0296023020
20 kg
14252.76 NOK
20kg
Please to purchase this item. Need a web account? Register with us today!
11467091
View Documents
MP Biomedicals
0296023050
50 kg
30979.56 NOK
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Research diet low in sodium (Na) and potassium (K).
Specifications

Specifications

Animal Diet
Single-use
Non-sterile
1 ft.
Autoclavable
Pellet
Room Temperature
0.3048 m
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.