Learn More
Cole-Parmer™ Polypropylene Two-Way Elliptic Ball Valves
426.00 NOK - 595.00 NOK
Specifications
Product Type | Two-Way Elliptic Ball Valve |
---|---|
Material | Polypropylene |
Min. Operating Temperature (Metric) | 110°C |
Packaging Quantity | 1 Pack |
Max. Operating Temperature (Metric) | 110°C |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11818073
|
Cole-Parmer™
06470-25 |
1/4 in. NPT (F) |
525.00 NOK
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11808073
|
Cole-Parmer™
06470-15 |
1/8 in. NPT (F) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11828073
|
Cole-Parmer™
06470-35 |
3/8 in. NPT(F) |
595.00 NOK
Each |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
Specifications
Two-Way Elliptic Ball Valve | |
110°C | |
110°C |
Polypropylene | |
1 Pack |