Learn More
Cole-Parmer™ Polypropylene Two-Way Elliptic Ball Valves
Brand: Cole-Parmer™ 06470-15
This item is not returnable.
View return policy
Specifications
Two-Way Elliptic Ball Valve | |
1/8 in. NPT (F) | |
1 Pack |
Polypropylene | |
110°C | |
110°C |