missing translation for 'onlineSavingsMsg'
Learn More

Polypeptide GalNac Transferase 7/GALNT7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

4250.00 NOK

Specifications

Antigen Polypeptide GalNac Transferase 7/GALNT7
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18329746
View Documents
Bio-Techne
NBP3-09941-100UL
100 μg
4250.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Polypeptide GalNac Transferase 7/GALNT7 Polyclonal specifically detects Polypeptide GalNac Transferase 7/GALNT7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Polypeptide GalNac Transferase 7/GALNT7
Western Blot
Unconjugated
Rabbit
Human
EC 2.4.1, EC 2.4.1.-, EC 2.4.1.37, EC 2.4.1.41, GalNAcT7, GalNAc-T7, N-acetylgalactosaminyltransferase 7, Polypeptide GalNAc transferase 7, polypeptide N-acetylgalactosaminyltransferase 7, pp-GaNTase 7, Protein-UDP acetylgalactosaminyltransferase 7, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 7, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 7 (GalNAc-T7)
The immunogen is a synthetic peptide directed towards the N-terminal region of Human Polypeptide GalNac Transferase 7/GALNT7. Peptide sequence SPLSRMREDRDVNDPMPNRGGNGLAPGEDRFKPVVPWPHVEGVEVDLESI
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
51809
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.