Learn More
Brand™ Polycarbonate Desiccators
Includes polycarbonate lid, polypropylene base and desiccant tray. BRAND™ Polycarbonate Desiccators feature a venting stopper with non-return valve in lid and polychlorobutadiene rubber (CK) O-ring compressed by vacuum to provide the seal between lid and base. Light-weight and easy to handle.
Brand:  Brand™ 65815
2373.32 NOK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 10212081
  • 2595.00 NOK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Includes polycarbonate lid, polypropylene base and desiccant tray.
  • Venting stopper with non-return valve in lid.
  • Polychlorobutadiene rubber (CK) O-ring, compressed by vacuum provides seal between lid and base.
  • Light-weight and easy to handle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polycarbonate Lid, Polypropylene Base and Dessicant Tray
Polychlorobutadiene rubber (CK) O-ring
285 mm
Desiccator
300 mm
Product Title
Brand™ Polycarbonate Desiccators > Size: 250mm