Learn More
HUBER Bath Cover
Bath covers, accessories for Huber temperature control units
Brand:  HUBER 6180
 View more versions of this product
Product Code. 10606527
  • 4130.00 NOK / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Description
Bath cover, insulated with opening
Specifications
Specifications
22 L Variostat Tank
Documents
One moment while we fetch your results.
Product Title
HUBER Bath Cover > For Use With: 22L Variostat Tank