Learn More
Eppendorf™ Pipet Helper™
The perfect instrument for inexperienced users as its robust and intuitive design allows everybody to adjust the meniscus precisely. In addition the valve unit is optimized so that liquids can be drawn up simply, without exerting pressure. The aspiration bulb provides rapid filling of the pipet.
Brand:  Eppendorf™ 4423000010
1113.81 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15230745
  • 1234.92 NOK / Each
In Stock
to see stock.
Explore more special offers
Description
  • Lightweight with reduced operating force
  • Easy maintenance and cleaning due to easy disassembly, few parts and fully autoclavable
  • Comfortable working because of the good balance and pleasing material
  • The aspirating bulb enables pipetting of volumes up to 100 mL at once
  • Single lever controls low force aspirating and dispensing
  • High control over the liquid movement due to very sensitive valve unit
  • Drop wise dispensing and blow out without splashing
  • Drainage without any additional pressure delivers very reproducible results
  • Drip stop via hydrophobic membrane filter (3 μm)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1
0.1 to 200 mL
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    5
    Hours
    17
    Minutes
    27
    Seconds
    54
Product Content Correction
Product Title
Eppendorf™ Pipet Helper™ >