Learn More
Eppendorf™ Easypet™ 3
Experience of a new dimension of electronic pipetting with this pipette controller. Eppendorf Easypet™ 3 offers intuitive and convenient speed adjustment. Ergonomic design allows fatigue-free pipetting.
Brand: Eppendorf™ 4430000018
6576.08 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Lightweight and well-balanced
- Vibrant backlit LEDs provide optical feedback of the remaining battery life
- Lithium polymer rechargeable battery offers long cordless runtime
- Operation while recharging is possible
- Autoclavable pipette adapter for sterile applications
- Quick release of aspirating cone for easy exchange of membrane filters
Specifications
Lithium-ion | |
Electronic Pipette | |
Membrane |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now