Learn More
BRAND™ Polypropylene Conical Ground Joint Stopper
Brand: BRAND™ 144430
175.48 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Conical bottom | |
NS 24/29 | |
Polypropylene | |
Square |
1/Ea. | |
Autoclavable | |
Ground Joint Stopper |
Product Content Correction
Product Title
BRAND™ Polypropylene Conical Ground Joint Stopper > Size: 24/29 NS