Learn More
Elma™ Pin Sets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
181.00 NOK - 600.00 NOK
Specifications
Quantity | 10 Pins/Set |
---|---|
Type | Pin Set |
For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | ||||||
17881307
|
Elma™
1125209 |
0509 |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17841367
|
Elma™
1125210 |
0158 |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17811317
|
Elma™
1125211 |
0767 |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17861367
|
Elma™
1125212 |
0613/0614 |
600.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17891347
|
Elma™
1125213 |
0772 |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17801317
|
Elma™
1125214 |
0741 |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17821357
|
Elma™
1125215 |
0001R |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
17821317
|
Elma™
1125216 |
0021R |
181.00 NOK
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
Description
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Specifications
10 Pins/Set | |
Pin Set | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Product Title
Elma™ Pin Sets