Learn More
Elma™ Pin Sets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
Brand:  Elma™ 1125214
 View more versions of this product
Product Code. 17801317
  • 181.00 NOK / Each
Only null left
Estimated Shipment: 07-07-2025
Description
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
10 Pins/Set
Pin Set
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
0741
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Elma™ Pin Sets > Pin Sets; Model: 0741