Learn More
Tocris Bioscience™ PF 5274857 hydrochloride
High affinity and selective Smoothened (Smo) receptor antagonist
Brand: Tocris Bioscience™ 5262/50
This item is not returnable.
View return policy
Description
PF 5274857 hydrochloride is a high affinity and selective Smoothened (Smo) receptor antagonist (Ki = 4.6 nM). Exhibits >1000-fold selectivity for Smo over a panel of other receptors, ion channels and enzymes. Inhibits Shh-induced Hedgehog signaling in MEF cells. Induces tumor regression in a medulloblastoma allograft mouse model. Orally available and brain penetrant.
Specifications
>98% | |
PF 5274857 hydrochloride | |
PF 5274857 hydrochloride |
50 mg | |
473.42 |
Product Content Correction
Product Title
Tocris Bioscience™ PF 5274857 hydrochloride > 50mg