Learn More
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
32.10 NOK - 448.00 NOK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
11797607
|
Fisherbrand™
18 08 0913 |
18 mm |
448.00 NOK
Pack of 1000 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
11797617
|
Fisherbrand™
22 08 0794 |
22 mm |
372.00 NOK
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
11510565
|
Fisherbrand™
22 08 0794 |
22 mm | N/A | |||||||
Description
- PE Snap Cap
- Transparent
- Closed top
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Transparent | |
Polyethylene | |
Snap Cap |
Product Title
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial