Learn More
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 22 08 0794
This item is not returnable.
View return policy
Description
- PE Snap Cap
- Transparent
- Closed top
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Transparent | |
Polyethylene | |
ND22 |
22 mm | |
Snap Cap |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial > 22mm