Learn More
MP Biomedicals Pantothenic Acid Control Diet, Pellet
Pantothenic acid control diet for rabbits. Low pantothenic acid diet 66 mk/kg.
Brand:  MP Biomedicals 0296021820
 View more versions of this product
Product Code. 11413011
  • 17920.00 NOK / 20kg
Description
  • Pantothenic acid control diet for rabbits
  • Low pantothenic acid diet 66 mg/kg
  • Contains casein, methionine, mineral mix and MP Bio vitamin fortification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Single-use
20 kg
Room Temperature
0.3048 m
Animal Diet
Pellet
Non-sterile
1 ft.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
MP Biomedicals Pantothenic Acid Control Diet, Pellet > Pantothenic Acid Control Diet, Pellet, 20 kg
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.