Learn More
Masterflex™ Opaque Black Viton 2-stopped Pump Tubing
Brand: Masterflex™ 97628-38
This item is not returnable.
View return policy
Specifications
2-stop Tubing | |
1.65 mm | |
30.48 cm | |
-13°C | |
Acids, Organic Solvents | |
Opaque Black |
Autoclavable | |
Viton | |
Acid and solvent transfer, high-temperature work applications | |
12 Pack | |
0.035 to 71mL/min. | |
-13°C to +204°C |