Learn More
Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes
Store tubes for automated picking/placing, liquid handling and other high throughput applications using Nunc Universal Latch Rack for Cryogenic Tubes.
Brand: Thermo Scientific™ 331828
1949.80 NOK valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes. Features a transparent cover with options for tubes of several sizes.
- Non-sterile
Ordering Information:
Case of 10
Specifications
Transparent | |
3.6 to 5 mL Cryogenic Tubes | |
Cryogenic Tube Storage Rack (w/Latch) |
For 3.6-5mL Standard Cryotubes, non-sterile | |
10/pk., 10/cs. |
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now