Learn More
Invitrogen™ UltraPure™ DEPC-Treated Water Product Code.: 11528616
Invitrogen™ UltraPure™ DEPC-Treated Water
Product Code. 11528616
4 x 100 mL, 400mL
Click to view available options
Quantity:
4 x 1.25 mL
1 L
4 x 100 mL
Unit Size:
1 L
400mL
Pack of 4
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Product Code. 11528616
Brand: Invitrogen™ 750024
Get 3 for 2
Use promo code 24112 at checkout
View Available Promotions
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Autoclaved to remove the DEPC and 0.1-ÎĽm membrane-filtered.
UltraPure™ DEPC-treated Water is suitable for use with RNA. It is prepared by incubating with 0.1% diethylpyrocarbonate (DEPC), and is then autoclaved to remove the DEPC. Sterile filtered.
Performance and Quality Testing: No DNase or RNase activity detected.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Grade Molecular Biology
Packaging Bottle
Treatment(s) DEPC-Treated
Quantity 4 x 100 mL
Purification Method Autoclaved, Membrane-Filtered
Shipping Condition Room Temperature
Product Line UltraPure™
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2930856 Certificate TypeCertificate of Analysis Date06/02/2025 Product Code750024
Lot Number2560667 Certificate TypeCertificate of Analysis Date06/02/2025 Product Code750024
Lot Number2848106 Certificate TypeCertificate of Analysis Date06/02/2025 Product Code750024
Lot Number2656134 Certificate TypeCertificate of Analysis Date06/02/2025 Product Code750024
Lot Number2538163 Certificate TypeCertificate of Analysis Date06/02/2025 Product Code750024
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ UltraPure™ DEPC-Treated Water >