Learn More
Ansell BioClean™ Durableu™ High Durability Cleanroom Overshoes Product Code.: 15228118
Ansell BioClean™ Durableu™ High Durability Cleanroom Overshoes
Product Code. 15228118
Size: 18 in.; Blue
Click to view available options
Size: 18 in.; Blue
Product Code. 15228118
Brand: Ansell BDBO18
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
BioClean Durableu™ disposable overshoes offer superb protection and durability. Resistant to tears, abrasions and a wide range of liquid chemicals, these cleanroom overshoes are low-linting, slip-resistant and have high tensile strength.
  • High Tensile Strength
  • Slip-Resistant
  • Resists tears and abrasions
  • Non-Woven spunbonded Polypropylene/cast polyethylene laminate
  • Manufactured in a facility holding ISO 9001:2008
  • In accordance with ISO 2859
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Material Polyethylene, Polypropylene
Color Blue
Certifications/Compliance ISO 9001:2015
Packaging Quantity 50 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; eight outer bags per lined carton (400 pieces)
  • Nitritex Bioclean-D
    Video Player is loading.
    Current Time 0:00
    Duration 3:12
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 3:12
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Nitritex Bioclean-D
Product Content Correction
Product Title
Ansell BioClean™ Durableu™ High Durability Cleanroom Overshoes >