Learn More
Nitritex™ BioClean-D Disposable Laboratory Coats
Wear Nitritex™ BioClean-D Disposable Laboratory Coats to protect against laboratory contamination.
Brand: Nitritex™ BDLC-L
1990.31 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Open cuff style
- Three pockets
- CleanTough™ Fabric
- Disposable, packaged in bag
- Spunbond Polypropylene
Specifications
Controlled Environments Lab Coat | |
Double Laminate Microporous Fabric | |
Non-sterile | |
ISO 9001:2015, ASTM F739-12 | |
Spunbond Polypropylene |
Large | |
Single-use | |
990.6 to 1066.8 mm | |
(EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2 |
Videos
-
Nitritex Bioclean-D
Product Content Correction
Product Title
Nitritex™ BioClean-D Disposable Laboratory Coats > Size: Large