missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE1/SLC9A1 Antibody (SR1132), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21872-50ul
This item is not returnable.
View return policy
Description
NHE1/SLC9A1 Monoclonal antibody specifically detects NHE1/SLC9A1 in Human samples. It is validated for Western Blot
Specifications
NHE1/SLC9A1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1132 | |
Western Blot 1:500-1:2000 | |
APNH1, APNHFLJ42224, Na(+)/H(+) antiporter, amiloride-sensitive, Na(+)/H(+) exchanger 1, Na+/H+, amiloride sensitive), Na-Li countertransporter, NHE-1, NHE1Na+/H+ antiporter, amiloride-sensitive, sodium/hydrogen exchanger 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 1 (antiporter, solute carrier family 9 (sodium/hydrogen exchanger), member 1, solute carrier family 9 (sodium/hydrogen exchanger), member 1 (antiporter, Solute carrier family 9 member 1 | |
A synthesized peptide derived from human NHE1/SLC9A1 (Uniprot #: P19634) | |
50 μg | |
Neuroscience | |
6548 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NHE1/SLC9A1 Antibody (SR1132), Novus Biologicals™ > 50 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction