Learn More
RETSCH Stainless Steel Grinding Jar
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand:  RETSCH 01.462.0331
 View more versions of this product
Product Code. 14821631
  • 25085.00 NOK / Each
Only null left
Estimated Shipment: 13-08-2025
Description
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Specifications
Grinding Jar
10 mL
For CryoMill
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Stainless Steel Grinding Jar > Capacity: 10 mL; For CryoMill