missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NET1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | NET1 |
---|---|
Concentration | 467 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16846035
|
Proteintech
28180-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16836035
|
Proteintech
28180-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NET1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NET1. This antibody recognizes human, mouse antigen. The NET1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
NET1 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG |
467 μg/mL | |
Polyclonal | |
Rabbit | |
10276 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title