Learn More
NET1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28180-1-AP-150UL
 View more versions of this product
Product Code. 16836035
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
The NET1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NET1. This antibody recognizes human, mouse antigen. The NET1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
NET1
Polyclonal
Unconjugated
150 μL
10276
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
467 μg/mL
Rabbit
Primary
Human, Mouse