missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, MP Biomedicals™
1192.50 NOK - 4319.50 NOK
Chemical Identifiers
CAS | 51298-62-5 |
---|---|
Molecular Formula | C7H16ClN5O4 |
Molecular Weight (g/mol) | 269.69 |
MDL Number | MFCD00039052,MFCD00133613 |
InChI Key | QBNXAGZYLSRPJK-JEDNCBNOSA-N |
Synonym | h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome |
PubChem CID | 135193 |
IUPAC Name | hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride |
SMILES | [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475051
|
MP Biomedicals
0215584605 |
5 g |
1192.50 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485051
|
MP Biomedicals
0215584610 |
10 g |
2130.60 NOK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495051
|
MP Biomedicals
0215584625 |
25 g |
4319.50 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
51298-62-5 | |
269.69 | |
QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
135193 | |
[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
C7H16ClN5O4 | |
MFCD00039052,MFCD00133613 | |
h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride |
Specifications
51298-62-5 | |
C7H16ClN5O4 | |
h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O | |
269.69 | |
269.7 | |
N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride |
White | |
MFCD00039052,MFCD00133613 | |
QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride | |
135193 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 257-116-1
Recommended Storage : Store at -0°C.