missing translation for 'onlineSavingsMsg'
Learn More

N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, MP Biomedicals™

Product Code. 11485051
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51298-62-5
Molecular Formula C7H16ClN5O4
Molecular Weight (g/mol) 269.69
MDL Number MFCD00039052,MFCD00133613
InChI Key QBNXAGZYLSRPJK-JEDNCBNOSA-N
Synonym h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome
PubChem CID 135193
IUPAC Name hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride
SMILES [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O

Specifications

Color White
Formula Weight 269.7
Quantity 10 g
Physical Form Powder
Chemical Name or Material N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'