missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate, 97%, Thermo Scientific Chemicals

940.00 NOK - 3615.00 NOK

Chemical Identifiers

CAS 82228-89-5
Molecular Formula C13H13NO4S
Molecular Weight (g/mol) 279.31
MDL Number MFCD00043164
InChI Key HSVLGIFAXFDLMU-UHFFFAOYSA-M
Synonym 4-formyl-1-methylpyridinium benzenesulfonate, 4-formyl-1-methyl-pyridinium benzenesulfonate, 4-formyl-1-methylpyridin-1-ium benzenesulfonate, pyridinium, 4-formyl-1-methyl-, benzenesulfonate, benzenesulfonate; 1-methylpyridin-1-ium-4-carbaldehyde, 4-formyl-1-methyl pyridinium benzensulfonate, 4-formyl-l-methyl pyridinium benzensulfonate, 4-formyl-1-methylpyridinium benzenesulfo nate, benzenesulfonic acid, 1-methylpyridine-4-carbaldehyde
PubChem CID 9817127
IUPAC Name benzenesulfonate;1-methylpyridin-1-ium-4-carbaldehyde
SMILES C[N+]1=CC=C(C=O)C=C1.[O-]S(=O)(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11308605
SDS View Documents
Thermo Scientific Alfa Aesar
L12072.06
5 g
940.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
11318605
SDS View Documents
Thermo Scientific Alfa Aesar
L12072.14
25 g
3615.00 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.

Solubility
Sparingly soluble in water (0.27 g/L at 25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under inert gas. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.