Learn More
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate, 97%, Thermo Scientific Chemicals
Description
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate is a useful reagent for the chemical transformation of primary amines to aldehydes and ketones under extremely mild conditions.
Solubility
Sparingly soluble in water (0.27 g/L at 25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under inert gas. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 98°C to 100°C |
Quantity | 5 g |
Beilstein | 5695963 |
Sensitivity | Air Sensitive |
Solubility Information | Sparingly soluble in water (0.27g/L at 25°C). |
Formula Weight | 279.32 (Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | N-Methylpyridinium-4-carboxaldehyde benzenesulfonate hydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10245568 | Certificate of Analysis | 06/02/2023 | L12072.14,L12072.06 |
10238123 | Certificate of Analysis | 12/04/2022 | L12072.14,L12072.06 |
10201948 | Certificate of Analysis | 06/12/2021 | L12072.14,L12072.06 |
10188029 | Certificate of Analysis | 06/12/2021 | L12072.14,L12072.06 |
10141101 | Certificate of Analysis | 06/12/2021 | L12072.14,L12072.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.