missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™

755.83 NOK - 3509.19 NOK

Chemical Identifiers

CAS 98064-96-1
Molecular Formula C28H54O11
Molecular Weight (g/mol) 566.729
MDL Number MFCD01310880
InChI Key GTQCHJYVKDXMRU-YMEALESQSA-N
Synonym n-hexadecyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cetyl, cetyl beta-d-maltoside, hexadecyl beta-maltoside, hexadecyl-, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl
PubChem CID 9850929
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15497929
SDS View Documents
Thermo Scientific Alfa Aesar
J66531.MC
100 mg N/A N/A N/A
15407939
SDS View Documents
Thermo Scientific Alfa Aesar
J66531.ME
500 mg N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C; Store under Argon

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.