missing translation for 'onlineSavingsMsg'
Learn More
n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™ Product Code.: 15407939

n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™

Product Code. 15407939
500 mg, 500mg
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15407939

Brand: Thermo Scientific Alfa Aesar J66531.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98064-96-1
Molecular Formula C28H54O11
Molecular Weight (g/mol) 566.729
MDL Number MFCD01310880
InChI Key GTQCHJYVKDXMRU-YMEALESQSA-N
Synonym n-hexadecyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cetyl, cetyl beta-d-maltoside, hexadecyl beta-maltoside, hexadecyl-, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl
PubChem CID 9850929
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Color White
Quantity 500 mg
Sensitivity Moisture sensitive
Formula Weight 566.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material n-Hexadecyl beta-D-maltopyranoside
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.