missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fat Diet, High Unsaturated Fat

9445.00 NOK - 29755.00 NOK

Specifications

Product Type Fat Diet
Components Casein Purified High Nitrogen 20%, DL-Methionine 0.3%, Sucrose 30.58%, Corn Starch 20%, Safflower Oil 20%, Alphacel, Non-Nutritive Bulk 5%, DL-a-Tocopherol Powder 0.12%, AIN-76 Mineral Mix 4%
Format Pellet
Storage Requirements 15°C to 30°C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11467131
View Documents
MP Biomedicals™
0296024410
10,000 g
9445.00 NOK
10kg
Please to purchase this item. Need a web account? Register with us today!
11487131
View Documents
MP Biomedicals™
0296024450
50,000 g
29755.00 NOK
50kg
Please to purchase this item. Need a web account? Register with us today!
11477131
View Documents
MP Biomedicals™
0296024420
20,000 g
13710.00 NOK
20kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Fat Diet
Casein Purified High Nitrogen 20%, DL-Methionine 0.3%, Sucrose 30.58%, Corn Starch 20%, Safflower Oil 20%, Alphacel, Non-Nutritive Bulk 5%, DL-a-Tocopherol Powder 0.12%, AIN-76 Mineral Mix 4%
Pellet
15°C to 30°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Fat Diet, High Unsaturated Fat

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.