missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fat Diet, High Unsaturated Fat
Brand: MP Biomedicals™ 0296024420
This item is not returnable.
View return policy
Description

Specifications
Fat Diet | |
Pellet | |
20,000 g |
Casein Purified High Nitrogen 20%, DL-Methionine 0.3%, Sucrose 30.58%, Corn Starch 20%, Safflower Oil 20%, Alphacel, Non-Nutritive Bulk 5%, DL-a-Tocopherol Powder 0.12%, AIN-76 Mineral Mix 4% | |
Powder | |
15°C to 30°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Fat Diet, High Unsaturated Fat > 20kg; Pelleted
Spot an opportunity for improvement?Share a Content Correction