Learn More
Heidolph instruments MonoBlock Flask Insert
Accommodate specifically sized flasks at your StarFish magnetic stirrer workstation. Heidolph™ MonoBlock Flask Insert, a one-piece MonoBlock insert for flasks, is ideal for experiments using the same vessel.
Brand: Heidolph instruments 505-80044-00
This item is not returnable.
View return policy
Description
- Accommodates one specific flask size.
- One-piece construction.
Specifications
Monoblock insert | |
Starfish Workstation, MR-Hei Magnetic Stirrers, 5 x 250 mL Flask MonoBlock | |
10 mL Flask |