missing translation for 'onlineSavingsMsg'
Learn More

MON1B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen MON1B
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17561336
View Documents
Proteintech
17638-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
17551336
View Documents
Proteintech
17638-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MON1B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MON1B. This antibody recognizes human antigen. The MON1B antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

MON1B
Polyclonal
Rabbit
22879
Primary
Western Blot, Immunofluorescence
Unconjugated
Human
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.