Learn More
MON1B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17638-1-AP-20UL
 View more versions of this product
Product Code. 17561336
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The MON1B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MON1B. This antibody recognizes human antigen. The MON1B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MON1B
Polyclonal
Rabbit
Primary
Human
Western Blot, Immunofluorescence
Unconjugated
20 μL
22879