missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%

CAS: 712313-64-9 | C8H16ClNO2 | 193.67 g/mol

Brand:  Thermo Scientific Alfa Aesar H62024.03

Quantity 1 g

 View more versions of this product

Product Code. 15436936

  • 7350.00 NOK / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC18Z005 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberH62024.MD,H62024.03
Lot NumberP23H027 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62024.MD,H62024.03
Lot NumberC18Z005 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62024.MD,H62024.03
Lot NumberM12E059 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62024.MD,H62024.03
Lot NumberP10E033 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62024.MD,H62024.03
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95% > 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.