missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%

Product Code. 15446936
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 712313-64-9 | C8H16ClNO2 | 193.67 g/mol

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 712313-64-9
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.67
MDL Number MFCD12022631,MFCD19237735,MFCD22392158,MFCD31620808,MFCD13184930,MFCD28167966
InChI Key OOFXENVWECZJBF-UHFFFAOYNA-N
Synonym trans-methyl-3-aminocyclohexanecarboxylate hydrochloride, trans-methyl-3-aminocyclohexanecarboxylatehydrochloride, methyl trans-3-aminocyclohexanecarboxylate hydrochloride, methyl 1r,3r-3-aminocyclohexane-1-carboxylate hydrochloride
PubChem CID 46172886
SMILES Cl.COC(=O)C1CCCC(N)C1

Specifications

Solubility Information Insoluble in water.
IUPAC Name methyl (1R,3R)-3-aminocyclohexane-1-carboxylate;hydrochloride
Quantity 250 mg
Formula Weight 193.67
Percent Purity 97%
Chemical Name or Material Methyl trans-3-aminocyclohexanecarboxylate hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.