Learn More
Brady™ MetaLabel™ B-486 Metallized Polyester Labels with an Aggressive Adhesive
For use in commercial/industrial applications. Brady B-486 metalized polyester labels with an aggressive adhesive are for use with BMP71 printer.
Brand:  Brady™ 114779
 View more versions of this product
Product Code. 11727104
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
UL Recognized, CSA Approved
Rubber, Permanent
Matte
Asset Tracking Label
BMP™71 Printer
2 x 1 in. (W x H)
0.0053 in.
Chemical, Corrosion, Humidity, UV and Weather-Resistant
Light Gray
Metallized Polyester (B-486)
Asset tracking, Rating plates, General purpose ID
100/Pk.
50.8 x 25.4 mm (W x H)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ MetaLabel™ B-486 Metallized Polyester Labels with an Aggressive Adhesive > W x H: 50.8 x 25.4mm (2 x 1 in.)