missing translation for 'onlineSavingsMsg'
Learn More

Melanophilin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen Melanophilin
Concentration 0.37 mg/mL
Applications Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886663
View Documents Promotion Details
Proteintech
10338-1-AP-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16876663
View Documents Promotion Details
Proteintech
10338-1-AP-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor protein MYO5A.
Specifications

Specifications

Melanophilin
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q9BV36
79083
Melanophilin Fusion Protein Ag0316
Primary
-20°C
MLPH
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Exophilin 3, melanophilin, MLPH, SLAC2 A, SLAC2A, Synaptotagmin like protein 2a
MLPH
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications