Learn More
Melanophilin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10338-1-AP-150UL
This item is not returnable.
View return policy
Description
Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor protein MYO5A.Specifications
Melanophilin | |
Polyclonal | |
Unconjugated | |
MLPH | |
Exophilin 3, melanophilin, MLPH, SLAC2 A, SLAC2A, Synaptotagmin like protein 2a | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79083 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BV36 | |
MLPH | |
Melanophilin Fusion Protein Ag0316 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |