Learn More
MC-DOXHZN hydrochloride, MedChemExpress
MedChemExpress MC-DOXHZN ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), with acid-sensitive properties.

POA - 4567.28 NOK
Specifications
Chemical Name or Material | MC-DOXHZN hydrochloride |
---|---|
CAS | 480998-12-7 |
Color | Reddish Brown |
Purity Grade Notes | Research |
Molecular Formula | C37H43ClN4O13 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16479557
![]() |
Medchem Express
HY-16261B |
5 mg |
2607.62 NOK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16433067
![]() |
Medchem Express
HY-16261B |
50 mg |
POA
50mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16427018
![]() |
Medchem Express
HY-16261B |
10 mg |
4567.28 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MC-DOXHZN ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), with acid-sensitive properties.Specifications
MC-DOXHZN hydrochloride | |
Reddish Brown | |
C37H43ClN4O13 | |
DMSO : 25 mg/mL (31.76 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (15.88 mM; ultrasonic and warming and heat to 60°C) | |
787.21 | |
98.0% | |
Solid |
480998-12-7 | |
Research | |
(E/Z)-Aldoxorubicin hydrochlorideDoxorubicin(6-maleimidocaproyl)hydrazone hydrochloride | |
[H][C@@]1(O[C@H]2C[C@@](/C(CO)=N\NC(CCCCCN3C(C=CC3=O)=O)=O)(O)CC(C2=C4O)=C(O)C5=C4C(C6=C(OC)C=CC=C6C5=O)=O)O[C@@H](C)[C@@H](O)[C@@H](N)C1.[H]Cl | |
787.21 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
missing translation for 'shelfLife' : -20°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Recommended Storage : -20°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.