Learn More
MC-DOXHZN hydrochloride, MedChemExpress
MedChemExpress MC-DOXHZN ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), with acid-sensitive properties.
Brand: Medchem Express HY-16261B

Description
MC-DOXHZN ((E/Z)-Aldoxorubicin) hydrochloride is an albumin-binding prodrug of Doxorubicin (DNA topoisomerase II inhibitor), with acid-sensitive properties.Specifications
MC-DOXHZN hydrochloride | |
Reddish Brown | |
C37H43ClN4O13 | |
(E/Z)-Aldoxorubicin hydrochlorideDoxorubicin(6-maleimidocaproyl)hydrazone hydrochloride | |
[H][C@@]1(O[C@H]2C[C@@](/C(CO)=N\NC(CCCCCN3C(C=CC3=O)=O)=O)(O)CC(C2=C4O)=C(O)C5=C4C(C6=C(OC)C=CC=C6C5=O)=O)O[C@@H](C)[C@@H](O)[C@@H](N)C1.[H]Cl | |
787.21 | |
Research | |
Cancer-programmed cell death |
480998-12-7 | |
Research | |
50 mg | |
DMSO : 25 mg/mL (31.76 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (15.88 mM; ultrasonic and warming and heat to 60°C) | |
787.21 | |
98.0% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only