missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Rabbit anti-Human, Mouse, Rat, CoraLite 555, Clone: 3O10, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL555-80137-20UL
This item is not returnable.
View return policy
Description
MAP2K3 antibody apecifically detetects MAP2K3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
MAP2K3 | |
3O10 | |
CoraLite 555 | |
BC032478 | |
Rabbit | |
Protein A purification | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunofluorescence | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
MAP2K3 | |
Peptide | |
20 μL | |
5606 | |
Store at -20°C. Avoid exposure to light. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction