missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K3 Rabbit anti-Human, Mouse, Rat, CoraLite 555, Clone: 3O10, Proteintech
Rabbit Recombinant Antibody
1696.32 NOK - 5264.45 NOK
Specifications
Antigen | MAP2K3 |
---|---|
Clone | 3O10 |
Applications | Immunofluorescence |
Conjugate | CoraLite 555 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17741369
|
Proteintech
CL555-80137-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17639916
|
Proteintech
CL555-80137-100UL |
100 μL |
5264.45 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP2K3 antibody apecifically detetects MAP2K3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
MAP2K3 | |
Immunofluorescence | |
Liquid | |
Human, Mouse, Rat | |
BC032478 | |
MAP2K3 | |
IgG | |
Protein A purification |
3O10 | |
CoraLite 555 | |
Rabbit | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
5606 | |
Peptide | |
Primary | |
Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title