Learn More
Hellma Macro Cuvettes, 110-QS (pair)
Highly pure Suprasil™ quartz glass cuvette with a PTFE stopper.
Brand: Hellma 110-5-PK-40
This item is not returnable.
View return policy
Description
- High-purity Suprasil™ quartz construction
- UV/VIS range
- PTFE stoppers included
Specifications
1750 μL | |
PTFE | |
Base Thickness: 1.5 mm | |
2/Pk | |
200 to 2500 nm | |
12.5 mm |
Suprasil Quartz | |
9.5 mm | |
PTFE Stoppers | |
5 mm | |
46 mm |