Learn More
Hellma Macro Cuvettes, 110-QS (pair)
Highly pure Suprasil™ quartz glass cuvette with a PTFE stopper.
Brand:  Hellma 110-1-PK-40
 View more versions of this product
Product Code. 15910421
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • High-purity Suprasil™ quartz construction
  • UV/VIS range
  • PTFE stoppers included
Specifications
Specifications
350 μL
PTFE
Base Thickness: 1.5 mm
2/Pk
200 to 2500 nm
12.5 mm
Suprasil Quartz
9.5 mm
PTFE Stoppers
1 mm
52 mm