missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ ProDoma Hybridoma Media
1048.13 NOK
Specifications
Content And Storage | 2° to 8°C |
---|---|
Without Additives | No Glutamine, No Phenol Red |
Quantity | 1 L |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11635110
|
Lonza™
BE02-029Q |
ProDoma 1 | N/A | ||||||
Description
The original hybridoma media for murine, human, and chimeric origin cells.
- Non-animal, protein-free media for your hybridoma needs.
- Scalable from small flasks to large-scale bioreactors.
- Hybridoma cell growth
- Monoclonal antibody production
Specifications
2° to 8°C | |
No Glutamine, No Phenol Red | |
1 L |